VWR®, Dissecting forceps
Curved, serrated, fine tips, 152 mm
Catalog # VWRU82027-406
Supplier: VWR Collection
Jump to:
Product Details & Documents
Dissecting forceps made of stainless steel. Available straight or curved and in different lengths.
Specifications
- Type:Curved, serrated, fine tips
- Length:152 mm
- Seq:13