Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, Dissecting forceps
VWR®, Dissecting forceps
Straight, very fine tips (tower tip), 108 mm
Catalog # VWRU82027-402
Supplier:   VWR Collection
VWR®, Dissecting forceps
Catalog # VWRU82027-402
Supplier:  VWR Collection
VWR®, Dissecting forceps
VWR®, Dissecting forceps
Straight, very fine tips (tower tip), 108 mm
Price
$26.91
Log in for availability
1 * 1 items
Product Details & Documents
Dissecting forceps with extra fine tips. Made from stainless steel.
Specifications
  • Type:
    Straight, very fine tips (tower tip)
  • Length:
    108 mm
  • Seq:
    01