Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, Laboratory Notebooks

VWR®, Laboratory Notebooks

Lab book, Blue, Ruled, English, A4, 120
Catalog # VWRU89005-136
Supplier:   VWR Collection
VWR®, Laboratory Notebooks
Catalog # VWRU89005-136
Supplier:  VWR Collection
VWR®, Laboratory Notebooks
VWR®, Laboratory Notebooks

Lab book, Blue, Ruled, English, A4, 120

Price
Inquire For Price
Log in for availability
1 * 10 items
Product Details & Documents

Imitation leather cover with coating, for durability in laboratory environments. Acid-free, archive safe paper (90 gsm). Includes laboratory notebook guidelines page, table of contents and user data page. Available in different language formats: English only, English-French, English-German, English-Spanish and English-Italian.

  • Pages are sewn in sections with round binding so that book lays flat
  • Scientific 5 mm grid or ruled format with date and witness signature blocks
  • Waterproof

Format: A4 (21×29,7 cm) or A5 (14,8×21 cm). A4 books available in three sizes: 72, 120 and 216 pages. A5 available with 96 pages.

Colour: blue

All books made in accordance with GLP, GMP and GCP guidelines.

Specifications
  • Paper size:
    A4
  • Type:
    Ruled
  • Version:
    English
  • Colour:
    Blue
  • Description:
    Lab book
  • No. of pages:
    120
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.