Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, Dissecting forceps
VWR®, Dissecting forceps
Straight, serrated, with fine tips, 114 mm
Catalog # VWRU82027-408
Supplier:   VWR Collection
VWR®, Dissecting forceps
Catalog # VWRU82027-408
Supplier:  VWR Collection
VWR®, Dissecting forceps
VWR®, Dissecting forceps
Straight, serrated, with fine tips, 114 mm
Price
Inquire For Price
Log in for availability
1 * 10 items
Product Details & Documents
Dissecting forceps made of stainless steel. Available straight or curved and in different lengths.
Specifications
  • Type:
    Straight, serrated, with fine tips
  • Length:
    114 mm
  • Seq:
    08