2-Propanol ≥99.0% USP, Multi-Compendial, J.T.Baker®
Glass bottle, amber, narrow mouth, 4 L
Supplier: Avantor
2-Propanol ≥99.0% USP, Multi-Compendial, J.T.Baker®
Catalog # 9037-03
Supplier: Avantor
CAS Number: 67-63-0
Jump to:
Product Details & Documents
Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
- CAS Number:67-63-0
- Formula:(CH₃)₂CHOH
- MDL Number:MFCD00011674
- Molecular Weight:60.1 g/mol
- Merck Index:13,05228
- Boiling Pt:82 °C (1013 hPa)
- Melting Pt:–89 °C
- Density:0.786 g/cm³ (20 °C)
- Flash Pt:12 °C
- Storage Temperature:Ambient
- EINECS:200-661-7
- UN:1219
- ADR:3,II
- REACH:01-2119457558-25
Documents
Specifications
Specification Test Results
- Bulk Pharmaceutical Chemical:Passes Test
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Specific Gravity at 25°/25°C:0.783 - 0.787
- USP - Refractive Index, n²ºD:1.376 - 1.378
- USP - Acidity:Passes Test
- USP - Nonvolatile Residue:≤0.005 %
- USP - Assay:≥99.0 %
- USP - Water (H₂O):≤0.5 %
- USP - Volatile Impurities - Ethyl ether:≤0.1 %
- USP - Volatile Impurities - Acetone:≤0.1 %
- USP - Volatile Impurities - Diisopropyl ether:≤0.1 %
- USP - Volatile Impurities - 1-Propanol:≤0.1 %
- USP - Volatile Impurities - 2-Butanol:≤0.1 %
- USP - Volatile Impurities - Total:≤1.0 %
- USP - Benzene:≤2 ppm
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- Appearance:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Benzene (C₆H₆):≤2 ppm
- EP/BP - Any peak other than 2-butanol:≤0.3 %
- EP/BP - Peroxide:Passes Test
- EP/BP - Nonvolatile Matter:≤20 ppm
- EP/BP - Water (H₂O):≤0.5 %
- EP/BP - UV Absorbance - 310 nm:≤0.01
- EP/BP - UV Absorbance - 290 nm:≤0.02
- EP/BP - UV Absorbance - 270 nm:≤0.03
- EP/BP - UV Absorbance - 250 nm:≤0.10
- EP/BP - UV Absorbance - 230 nm:≤0.30
- EP/BP - Spectrum Appearance:Passes Test
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Specific Gravity at 20°/20°C:0.785 - 0.788
- JP - Clarity of Solution:Passes Test
- JP - Acidity:Passes Test
- JP - Residue on Evaporation, mg/20mL:≤1.0
- JP - Water (H₂O):≤0.75 %
- JP - Distilling Range(Lower):81 - 83 °C
- JP - Distilling Range (Upper):81 - 83 °C
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:4 L
- Cat. No.:9037-03
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



