2-Propanol ≥99.0% USP, Multi-Compendial, J.T.Baker®
Glass bottle, 500 mL
: Avantor
2-Propanol ≥99.0% USP, Multi-Compendial, J.T.Baker®
9037-01
: Avantor
: 67-63-0
: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
- :67-63-0
- :(CH₃)₂CHOH
- :MFCD00011674
- :60.1 g/mol
- :13,05228
- :82 °C (1013 hPa)
- :–89 °C
- :0.786 g/cm³ (20 °C)
- :12 °C
- :Ambient
- :200-661-7
- :1219
- :3,II
- :01-2119457558-25
Documents
- Bulk Pharmaceutical Chemical:Passes Test
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Specific Gravity at 25°/25°C:0.783 - 0.787
- USP - Refractive Index, n²ºD:1.376 - 1.378
- USP - Acidity:Passes Test
- USP - Nonvolatile Residue:≤0.005 %
- USP - Assay:≥99.0 %
- USP - Water (H₂O):≤0.5 %
- USP - Volatile Impurities - Ethyl ether:≤0.1 %
- USP - Volatile Impurities - Acetone:≤0.1 %
- USP - Volatile Impurities - Diisopropyl ether:≤0.1 %
- USP - Volatile Impurities - 1-Propanol:≤0.1 %
- USP - Volatile Impurities - 2-Butanol:≤0.1 %
- USP - Volatile Impurities - Total:≤1.0 %
- USP - Benzene:≤2 ppm
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- Appearance:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Benzene (C₆H₆):≤2 ppm
- EP/BP - Any peak other than 2-butanol:≤0.3 %
- EP/BP - Peroxide:Passes Test
- EP/BP - Nonvolatile Matter:≤20 ppm
- EP/BP - Water (H₂O):≤0.5 %
- EP/BP - UV Absorbance - 310 nm:≤0.01
- EP/BP - UV Absorbance - 290 nm:≤0.02
- EP/BP - UV Absorbance - 270 nm:≤0.03
- EP/BP - UV Absorbance - 250 nm:≤0.10
- EP/BP - UV Absorbance - 230 nm:≤0.30
- EP/BP - Spectrum Appearance:Passes Test
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Specific Gravity at 20°/20°C:0.785 - 0.788
- JP - Clarity of Solution:Passes Test
- JP - Acidity:Passes Test
- JP - Residue on Evaporation, mg/20mL:≤1.0
- JP - Water (H₂O):≤0.75 %
- JP - Distilling Range(Lower):81 - 83 °C
- JP - Distilling Range (Upper):81 - 83 °C
- Pack type:Glass bottle
- Pk:500 mL
- Cat. No.:9037-01
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health



