Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Japan
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
XLT4 AGAR PETRI 90MM - 1 box with 2 packs of 10 plates/pack. Single cellophane. 1 * 20 items
Catalog # 101874ZA
Supplier:
VWR Chemicals
SDS
Certificates
Expand image
XLT4 AGAR PETRI 90MM - 1 box with 2 packs of 10 plates/pack. Single cellophane. 1 * 20 items
Catalog # 101874ZA
Supplier:
VWR Chemicals
SDS
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
101874ZA_XLT4 AGAR PETRI 90MM_ISO 11133_rev1
SDS
sds_VWRC101874ZA
Product specification
spec_VWRC101874ZA
Product Literature
VWR Microbiology
VWR® for Microbiology Enumeration Isolation Detection and Cultivation of Bacteria Yeast and Moulds
VWR® for Microbiology media
Documentation
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection