Order Entry
Japan
Orders LinkContactUsLinkComponent
2-Propanol ≥98%, TECHNICAL

2-Propanol ≥98%, TECHNICAL

Plastic drum, 25 L
Catalog # 20922.466
Flammable
Irritant
   Safer Human and Environmental Health
Supplier:   VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Catalog # 20922.466
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  67-63-0
2-Propanol ≥98%, TECHNICAL
2-Propanol ≥98%, TECHNICAL

Plastic drum, 25 L

CAS Number:  67-63-0
Price
Inquire For Price
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
  • CAS Number:67-63-0
  • Formula:(CH₃)₂CHOH
  • MDL Number:MFCD00011674
  • Molecular Weight:60.1 g/mol
  • Merck Index:13,05228
  • Boiling Pt:82 °C (1013 hPa)
  • Melting Pt:–89 °C
  • Density:0.786 g/cm³ (20 °C)
  • Flash Pt:12 °C
  • Storage Temperature:Ambient
  • EINECS:200-661-7
  • UN:1219
  • ADR:3,II
  • REACH:01-2119457558-25
Specifications

Specification Test Results

  • Assay:
    Min. 98 %

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    25 L
  • Cat. No.:
    20922.466
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.