Order Entry
Japan
Orders LinkContactUsLinkComponent
D-(-)-Fructose Ph. Eur.

D-(-)-Fructose Ph. Eur.

Plastic bucket, 25 kg
Catalog # 24282.461
Supplier:   VWR Chemicals
D-(-)-Fructose Ph. Eur.
Catalog # 24282.461
Supplier:  VWR Chemicals
CAS Number:  57-48-7
D-(-)-Fructose Ph. Eur.
D-(-)-Fructose Ph. Eur.

Plastic bucket, 25 kg

CAS Number:  57-48-7
Price
Inquire For Price
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
  • CAS Number:57-48-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148910
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04295
  • Boiling Pt:552 °C (1013 hPa)
  • Melting Pt:100…110 °C
  • Density:1.59 g/cm³ (20 °C. OECD 109)
  • Storage Temperature:Ambient
  • EINECS:200-333-3
Specifications

Specification Test Results

  • Appearance:
    Colourless crystals
  • Identification A:
    Passes test
  • Identification B:
    Passes test
  • Identification C:
    Passes test
  • Identification D:
    Passes test
  • Appearance of solution:
    Passes test
  • Acidity or alkalinity:
    Passes test
  • Spec. opt. rotation (calc. on anhydrous):
    -93.5 to -91 deg
  • Foreign sugars:
    Passes test
  • 5-Hydroxymethylfurfural + related subst.:
    Passes test
  • Ba (Barium):
    Passes test
  • Lead in sugars:
    Max. 0.5 ppm
  • Water:
    Max. 0.5 %
  • Sulphated ash:
    Max. 0.1 %
  • Residual solvents:
    Passes test

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    25 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.