Order Entry
Japan
Orders LinkContactUsLinkComponent
D-(-)-Fructose Ph. Eur.

D-(-)-Fructose Ph. Eur.

Plastic bottle for solids, 5 kg
Catalog # 24282.368
Supplier:   VWR Chemicals
D-(-)-Fructose Ph. Eur.
Catalog # 24282.368
Supplier:  VWR Chemicals
CAS Number:  57-48-7
D-(-)-Fructose Ph. Eur.
D-(-)-Fructose Ph. Eur.

Plastic bottle for solids, 5 kg

CAS Number:  57-48-7
Price
Inquire For Price
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
  • CAS Number:57-48-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148910
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04295
  • Boiling Pt:552 °C (1013 hPa)
  • Melting Pt:100…110 °C
  • Density:1.59 g/cm³ (20 °C. OECD 109)
  • Storage Temperature:Ambient
  • EINECS:200-333-3
Specifications

Specification Test Results

  • Appearance:
    Colourless crystals
  • Identification A:
    Passes test
  • Identification B:
    Passes test
  • Identification C:
    Passes test
  • Identification D:
    Passes test
  • Appearance of solution:
    Passes test
  • Acidity or alkalinity:
    Passes test
  • Spec. opt. rotation (calc. on anhydrous):
    -93.5 to -91 deg
  • Foreign sugars:
    Passes test
  • 5-Hydroxymethylfurfural + related subst.:
    Passes test
  • Ba (Barium):
    Passes test
  • Lead in sugars:
    Max. 0.5 ppm
  • Water:
    Max. 0.5 %
  • Sulphated ash:
    Max. 0.1 %
  • Residual solvents:
    Passes test

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    5 kg

Specifications