Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR® Sticky Mat

VWR® Sticky Mat

Sticky mat, 22×18 cm
Catalog # 444-1417
Supplier:   VWR Collection
VWR® Sticky Mat
Catalog # 444-1417
Supplier:  VWR Collection
VWR® Sticky Mat
VWR® Sticky Mat

Sticky mat, 22×18 cm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Easily secure and remove samples without clamps.

  • Recommemded maximum speed not to exceed 250 min⁻¹

  • Application temperature not to exceed 65 °C
  • Clean with water and air dry to restore adhesive quality

Caution: Recommended maximum speed should not exceed 250 rpm. Application tempeartures should not exceed 65 °C. Clean with water and air dry to restore adhesive qualities.

Specifications
  • Description:
    Sticky mat, 22×18 cm
  • Seq:
    0233
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.