Cell scrapers and cell lifters
Supplier: VWR Collection
About this item
Specifically designed to make collecting cells easier and more effective. Available with a choice of blade positions: scraper for use in flasks, or lifter for use in harvesting cells (especially stem cells) in dishes.
- Cross-ribbed handle in ABS provides greater rigidity and ensures better control while scraping cells
- Thin, flexible TPE blade prevents damage to cells
- Individually wrapped
- Sterilised by electron beam irradiation
- Free from DNase, RNase and non-pyrogenic
4 Options Available Below
Product Details & Documents
Specifically designed to make collecting cells easier and more effective. Available with a choice of blade positions: scraper for use in flasks, or lifter for use in harvesting cells (especially stem cells) in dishes.
- Cross-ribbed handle in ABS provides greater rigidity and ensures better control while scraping cells
- Thin, flexible TPE blade prevents damage to cells
- Individually wrapped
- Sterilised by electron beam irradiation
- Free from DNase, RNase and non-pyrogenic
Documents
Specifications for Product Family
Specifications
- Catalog No:
- 734-2602
- 734-2603
- 734-2604
- 734-2605
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
Specifications
Product Family Options
Product Information
- Total lengthAvailabilityPrice
- 734-2602Cell scraper, blade width 20 mmSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2602Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-2603Cell lifter, blade width 20 mmSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2603Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-2604Cell scraper, blade width 30 mmSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2604Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 734-2605Cell lifter, blade width 30 mmSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-2605Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact