VWR®, Horseradish peroxidase
Catalog # 0343-25000U
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Amber Bottle
- Pk:25 KU
- CAS No.:9003-99-0
- Species:Horseradish
- Storage conditions:Freezer
- Enzyme name:Peroxidase
- Enzyme activity:250 Ppg U/mg
- Tested applications:For biochemistry
Specifications
About this item
Catalyses the decomposition of hydrogen peroxide in solution causing the oxidation of a number of substrates. Used extensively as a label for immunohistological assays and ELISA. Its exceptional stability in powdered and solution form enhances its desirability as an enzyme conjugate.