Order Entry
Japan
ContactUsLinkComponent
MOPSO (3-(4-morpholino)-2-hydroxypropanesulphonic acid) free acid ≥98%, Ultra Pure Grade
Irritant
MOPSO (3-(4-morpholino)-2-hydroxypropanesulphonic acid) free acid ≥98%, Ultra Pure Grade
Catalog # J589-100G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
MOPSO (3-(4-morpholino)-2-hydroxypropanesulphonic acid) free acid ≥98%, Ultra Pure Grade
Catalog # J589-100G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Melting Point:
    276 – 284 °C
  • Moisture (KF):
    <1%
  • Purity:
    >98%
  • Solubility (33.3%, water):
    Pass

Specification Test Results

About this item

 :
 :225.27 g/mol
EINECS:269-989-6
 :GHS07
Storage Temperature:Ambient
CAS Number:68399-77-9
Formula:C₇H₁₅NO₅S
MDL Number:MFCD00009629