Order Entry
Japan
ContactUsLinkComponent
Iron(III) sulphate hydrate, Reagent Grade
Irritant
Iron(III) sulphate hydrate, Reagent Grade
Catalog # 0629-500G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Iron(III) sulphate hydrate, Reagent Grade
Catalog # 0629-500G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :399.88 g/mol (anhy)
Merck Index:13,04065
Density:3.1 g/cm³ (20 °C)
EINECS:233-072-9
 :GHS07
Storage Temperature:Ambient
CAS Number:15244-10-7
Formula:Fe₂(SO₄)₃·xH₂O
MDL Number:MFCD00149714