Order Entry
Japan
ContactUsLinkComponent
Ethyl acetate ≥99.6%, UltimAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Flammable
Irritant
Ethyl acetate ≥99.6%, UltimAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # V553-10
   Safer Human and Environmental Health
CAS Number:  
undefined
Flammable
Irritant
Ethyl acetate ≥99.6%, UltimAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # V553-10
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pk:
    4 L
  • Seq:
    5162
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • ACS - Assay (CH₃COOCH₂CH₃) (by GC, corrected for water):
    ≥99.6 %
  • ACS - Color (APHA):
    ≤10
  • ACS - Residue after Evaporation:
    ≤1 ppm
  • ACS - Titrable Acid (µeq/g):
    ≤0.8
  • ACS - Water (H₂O)(by KF, coulometric):
    ≤0.04 %
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 255 nm:
    ≤1.00
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 257 nm:
    ≤0.50
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 263 nm:
    ≤0.10
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 265 nm:
    ≤0.05
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 275 nm:
    ≤0.05
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:
    ≤0.02
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 330 - 400 nm:
    ≤0.01
  • Substances Darkened by H₂SO₄:
    Passes Test
  • Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:
    ≤1.0 ppb
  • ECD Sensitive Impurities (as Heptachl or Epoxide) Single Peak (ng/L):
    ≤10 ppb
  • FID Sensitive Impurities (as 2-Octano l) Single Peak (µg/L):
    ≤5 ppb

Specification Test Results

About this item

 :
 :88.11 g/mol
Merck Index:13,03792
Boiling Pt:77.1 °C (1013 hPa)
Melting Pt:–83 °C
Density:0.902 g/cm³ (20 °C)
Flash Pt:–4 °C
Storage Temperature:Ambient
EINECS:205-500-4
UN:1173
ADR:3,II
REACH:01-2119475103-46
 :GHS02
 :GHS07
CAS Number:141-78-6
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171