Order Entry
Japan
ContactUsLinkComponent
Acetone, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Flammable
Irritant
Acetone, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Acetone, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    Min 99.5 %
  • ACS - Assay (CH₃COCH₃) (by GC, corrected for water):
    ≥99.5 %
  • ACS - Aldehydes (as HCHO):
    ≤0.002 %
  • ACS - Color (APHA):
    ≤10
  • ACS - Isopropyl Alcohol (CH₃CHOHCH₃)(by GC):
    ≤0.05 %
  • ACS - Methanol (CH₃OH)(by GC):
    ≤0.05 %
  • ACS - Residue after Evaporation:
    ≤0.0005 %
  • ACS - Solubility in H₂O:
    Passes Test
  • ACS - Substances Reducing Permanganate:
    Passes Test
  • ACS - Titrable Acid (meq/g):
    ≤0.0003
  • ACS - Titrable Base (meq/g):
    ≤0.0006
  • ACS - Water (H₂O):
    ≤0.5 %
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 330 nm:
    ≤1.00
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 340 nm:
    ≤0.10
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 375 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 nm:
    ≤0.010

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together