Order Entry
Japan
ContactUsLinkComponent
Dibutylamine ≥99% (by GC), BAKER ANALYZED®, J.T.Baker®
Flammable
Corrosive
Toxic
Dibutylamine ≥99% (by GC), BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Flammable
Corrosive
Toxic
Dibutylamine ≥99% (by GC), BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99 %
  • Assay (C₈H₁₉N) (by GC):
    ≥99 %
  • Refractive Index, n²ºD:
    1.415 - 1.419

Specification Test Results

Related Information

Frequently Bought Together