

N,N-Dimethylacetamide ≥99.0% (by GC), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # 5407-08
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:≥99.0%
- ACS - Assay (CH₃CON(CH₃)₂) (by GC):≥99.0 %
- ACS - Residue after Evaporation:≤5 ppm
- ACS - Water (H₂O)(by Karl Fischer titrn):≤0.03 %
- Optical Absorbance (1-cm path vs water) - 270 nm:≤1.00
- Optical Absorbance (1-cm path vs water) - 280 nm:≤0.30
- Optical Absorbance (1-cm path vs water) - 290 nm:≤0.15
- Optical Absorbance (1-cm path vs water) - 310 nm:≤0.05
- Optical Absorbance (1-cm path vs water) - 320 nm:≤0.03
- Optical Absorbance (1-cm path vs water) - 360 - 400 nm:≤0.01
Specification Test Results
About this item
:
:87.12 g/mol
Merck Index:13,03254
Boiling Pt:166.1 °C (1013 hPa)
Melting Pt:–20 °C
Density:0.937 g/cm³ (20 °C)
Flash Pt:70 °C
EINECS:204-826-4
UN:2810
ADR:6.1,III
:Filtered through a 0.2 µm filter.
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:127-19-5
Formula:CH₃CON(CH₃)₂
MDL Number:MFCD00008686