Order Entry
Japan
ContactUsLinkComponent
VWR®, Drosophila Vials
VWR®, Drosophila Vials
Catalog # 734-1255
Supplier:  VWR Collection
undefined
VWR®, Drosophila Vials
Catalog # 734-1255
Supplier:  VWR Collection
Specifications
  • Cap included:
    No
  • Ø×H:
    28,5×95 mm
  • Packed:
    Bulk
  • Description:
    Wide
  • Seq:
    01
  • Pk:
    500
Specifications
About this item
These PS vials have a flat bottom straight wall design for convenient racking, stacking and filling.
The tray packed 10×10 row configuration is great for quick, effortless filling.
Bulk packed vials are layered with open-end orientation for convenient handling and smaller case size.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Trays and dividers
Product Information