Order Entry
Japan
ContactUsLinkComponent
 
 
 
Pyridine, dehydrated (max. 0.01% H₂O) ≥99.5%, AnalaR NORMAPUR® analytical reagent
  27197.295
 :  
undefined
 
 
Pyridine, dehydrated (max. 0.01% H₂O) ≥99.5%, AnalaR NORMAPUR® analytical reagent
  27197.295
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Appearance:
    Clear colourless liquid
  • Assay (calculated on anhydrous):
    Min. 99.5 %
  • Acidity or alkalinity:
    Max. 0.002 meq/g
  • Water (K.F.):
    Max. 0.0100 %
  • Residue on evaporation:
    Max. 0.0010 %
  • Al (Aluminium):
    Max. 0.5 ppm
  • B (Boron):
    Max. 0.02 ppm
  • Ba (Barium):
    Max. 0.1 ppm
  • Ca (Calcium):
    Max. 0.5 ppm
  • Cd (Cadmium):
    Max. 0.05 ppm
  • Co (Cobalt):
    Max. 0.02 ppm
  • Cr (Chromium):
    Max. 0.02 ppm
  • Cu (Copper):
    Max. 0.02 ppm
  • Fe (Iron):
    Max. 0.1 ppm
  • Mg (Magnesium):
    Max. 0.1 ppm
  • Mn (Manganese):
    Max. 0.02 ppm
  • Ni (Nickel):
    Max. 0.02 ppm
  • Pb (Lead):
    Max. 0.1 ppm
  • Sn (Tin):
    Max. 0.1 ppm
  • Zn (Zinc):
    Max. 0.1 ppm

 

 

 :110-86-1
 :
 :C₅H₅N
 :MFCD00011732
 :13,08060
 :115 °C (1013 hPa)
 :–44 °C
 :0.983 g/cm³ (20 °C)
 :17 °C
 :1282
 :3,II
 :79.1 g/mol
 :203-809-9
 :GHS02
 :GHS07
 :Ambient