Order Entry
Japan
ContactUsLinkComponent
Lanthanum(III) oxide ≥99.5%, GPR RECTAPUR®
Lanthanum(III) oxide ≥99.5%, GPR RECTAPUR®
Catalog # 24960.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
Lanthanum(III) oxide ≥99.5%, GPR RECTAPUR®
Catalog # 24960.260
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    Min. 99.5 %

Specification Test Results

About this item

 :
 :325.81 g/mol
Boiling Pt:4200 °C (1013 hPa)
Melting Pt:2315 °C
Density:6.51 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-200-5
CAS Number:1312-81-8
Formula:La₂O₃
MDL Number:MFCD00011071