
Diphosphorus pentoxide ≥98.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1314-56-3
:
Formula:P₂O₅
MDL Number:MFCD00011440
:141.94 g/mol
Merck Index:13,07441
Melting Pt:562 °C
Density:2.253 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-236-1
UN:1807
ADR:8,II
:GHS05
Specification Test Results
- Assay:Min. 98.0 %
- Insolubility in water:Max. 50 ppm
- Reducing substances (as P2O3):Max. 0.02 %
- Total N (Nitrogen):Max. 10 ppm
- Cl (Chloride):Max. 10 ppm
- SO4 (Sulphate):Max. 50 ppm
- As (Arsenic):Max. 150 ppm
- Cd (Cadmium):Max. 5 ppm
- Cu (Copper):Max. 20 ppm
- Fe (Iron):Max. 10 ppm
- K (Potassium):Max. 100 ppm
- Mn (Manganese):Max. 5 ppm
- Na (Sodium):Max. 0.02 %
- Ni (Nickel):Max. 20 ppm
- Pb (Lead):Max. 10 ppm
- Zn (Zinc):Max. 20 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number2,5L Plastic bottle for solids DIN 80 (ZPACK13252500) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
spec_VWRC21411.230 spec_VWRC21411.296
Product specification
VWR® for Drying and Filtration
Product Literature
Frequently Bought Together