Order Entry
Japan
ContactUsLinkComponent
 
 
Diphosphorus pentoxide ≥98.0%, GPR RECTAPUR®
  21410.293
 :  
undefined
 
Diphosphorus pentoxide ≥98.0%, GPR RECTAPUR®
  21410.293
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

 

 

  • Assay:
    Min. 98.0%
  • Heavy metals (as Pb):
    Max. 100 ppm
  • Reducing substances (as P₂O₃):
    Max. 0.02 %
  • Cl (Chloride):
    Max. 50 ppm

 

 

 :
 :141.94 g/mol
 :13,07441
 :562 °C
 :2.253 g/cm³ (20 °C)
 :Ambient
 :215-236-1
 :1807
 :8,II
 :GHS05
 :1314-56-3
 :P₂O₅
 :MFCD00011440