
Potassium tellurite, TECHNICAL
Catalog # 27074.154
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:50 g
- Seq:3368
Specifications
Specification Test Results
- Identification:Passes test
Specification Test Results
About this item
:
:253.79 g/mol
Merck Index:12,07850
Melting Pt:460…470 °C
Storage Temperature:Refrigerator
EINECS:232-213-1
UN:3284
ADR:6.1,II
:GHS06
CAS Number:7790-58-1
Formula:K₂TeO₃
MDL Number:MFCD00011379