Water, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7732-18-5
:
Formula:H₂O
MDL Number:MFCD00011332
:18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
:Management of Change (MOC) category = NI
Storage Temperature:Ambient
EINECS:231-791-2
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 0.0010
- UV Absorbance on Gradient Elution - Largest Peak at 254nm (au):≤0.001
- Information Only:= 0.1 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.1 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤0.2 ppb
- Filtration Test:Passes Test
- pH, For Information Only:= 2.0 ppm
- Residue after Evaporation:≤2.0 ppm
Specification Test Results
Related Information
Frequently Bought Together