Water for biotechnology nuclease-free, 0.2 µm filtered
:7732-18-5
:
:H₂O
:MFCD00011332
:18.02 g/mol
:13,10098
:100 °C (1013 hPa)
:0 °C
:1 g/cm³ (4 °C)
:Filtered through a 0.2 µm filter.
:Ambient
:231-791-2
- DNase:NONE
- Protease:NONE
- RNase:NONE
- Sterility:PASS
Frequently Bought Together