Order Entry
Japan
ContactUsLinkComponent
Lithium carbonate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Irritant
Lithium carbonate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Catalog # 25007.230
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Lithium carbonate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Catalog # 25007.230
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Insolubility in hydrochloric acid:
    Max. 100 ppm
  • Cl (Chloride):
    Max. 20 ppm
  • NH4 (Ammonium):
    Max. 5 ppm
  • NO3 (Nitrate):
    Max. 5 ppm
  • SO4 (Sulphate):
    Max. 0.02 %
  • Ca (Calcium):
    Max. 50 ppm
  • Fe (Iron):
    Max. 10 ppm
  • K (Potassium):
    Max. 50 ppm
  • Mg (Magnesium):
    Max. 10 ppm
  • Na (Sodium):
    Max. 50 ppm
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

About this item

 :
 :73.89 g/mol
Merck Index:13,05549
Melting Pt:720 °C
Density:2.11 g/cm³ (20 °C)
EINECS:209-062-5
 :GHS07
Storage Temperature:Ambient
CAS Number:554-13-2
Formula:Li₂CO₃
MDL Number:MFCD00011084