


Potassium bromate ≥99.8% (dry basis), AnalaR NORMAPUR® analytical reagent
Catalog # 26703.232
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Assay (dried substance):Min. 99.8 %
- Heavy metals (as Pb):Max. 5 ppm
- Insolubility in water:Max. 50 ppm
- Total N (Nitrogen):Max. 10 ppm
- Br (Bromide):Max. 0.02 %
- SO4 (Sulphate):Max. 50 ppm
- Fe (Iron):Max. 1 ppm
- Na (Sodium):Max. 10 ppm
Specification Test Results
About this item
:
:167 g/mol
Merck Index:12,07779
Melting Pt:350 °C
Density:3.218 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-829-8
UN:1484
ADR:5.1,II
:GHS03
:GHS08
:GHS06
CAS Number:7758-01-2
Formula:KBrO₃
MDL Number:MFCD00011359