VWR® Vacuum Desiccators
Supplier: VWR Collection
Jump to:
About this item
Borosilicate 3.3 glass.
- Precisely ground flat flanges and interchangeable tubulated lids
- Standard ground joint NS 24/29 junction tube in the lid, complete with stopcock plus PTFE spindle
- Base and lid with flat flange ground to fit
- High mechanical, thermal and chemical resistance
Due to the increased wall thickness, these desiccators can be used up to the maximum vacuum that is technically possible.
Certifications: DIN 12491
5 Options Available Below
Product Details & Documents
Borosilicate 3.3 glass.
- Precisely ground flat flanges and interchangeable tubulated lids
- Standard ground joint NS 24/29 junction tube in the lid, complete with stopcock plus PTFE spindle
- Base and lid with flat flange ground to fit
- High mechanical, thermal and chemical resistance
Due to the increased wall thickness, these desiccators can be used up to the maximum vacuum that is technically possible.
Certifications: DIN 12491
Documents
Product Family Options
vacuum desiccators
Product Information
- TypeNominal sizeØ ext.×HØ int.AvailabilityPrice
Spare bases for VWR® vacuum desiccators
Product Information
- Catalog #Usable heightNominal sizeDescriptionØ ext.×HAvailabilityPrice
Spare lids for VWR® vacuum desiccators
Product Information
- Catalog #Nominal sizeDescriptionTypeAvailabilityPrice
Spare desiccator plates for VWR® vacuum desiccators
Product Information
- Catalog #ØTypeAvailabilityPrice