Order Entry
Japan
ContactUsLinkComponent
Sodium dodecyl sulphate (SDS) ≥99.0%, Proteomics Grade
 
 
 
Sodium dodecyl sulphate (SDS) ≥99.0%, Proteomics Grade
  M107-250G
 :  
undefined
 
 
 
Sodium dodecyl sulphate (SDS) ≥99.0%, Proteomics Grade
  M107-250G
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

 

 

  • Abs.@260 nm (3%, Water):
    0.1
  • Abs.@280 nm (3%, Water):
    0.1
  • Chloride:
    0.1%
  • Copper:
    0.0005%
  • DNase:
    NONE
  • Heavy Metals (as Pb):
    0.0005%
  • Identification:
    NONE
  • Iron:
    0.0001%
  • Phosphate:
    0.0005%
  • Protease:
    NONE
  • Purity:
    99.0%
  • RNase:
    NONE
  • Solubility (10%, Water):
    PASS

 

 

 :
 :288.38 g/mol
 :14,08636
 :204…207 °C
 :1.1 g/cm³ (20 °C)
 :>100 °C
 :Ambient
 :205-788-1
 :1325
 :4.1,III
 :01-2119489461-32
 :GHS02
 :GHS07
 :GHS05
 :151-21-3
 :CH₃(CH₂)₁₁OSO₃Na
 :MFCD00036175