


Chloroform ≥99.8% stabilised for biotechnology
Catalog # 0757-500ML
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Abs.@ 245nm:≤1
- Abs.@ 255nm:≤0.25
- Abs.@ 260nm:≤0.15
- Abs.@ 270nm:≤0.05
- Abs.@ 290nm:≤0.01
- Abs.@ 400nm:<0.01
- Chloride:<10 ppm
- Color (APHA):<10
- Lead:<0.05 ppm
- Moisture (KF):<0.05%
- Purity:>99.8%
- Residue After Evaporation:<0.001%
- Titratable Free Acid:<0.5 meq/g
Specification Test Results
About this item
:
:119.38 g/mol
Merck Index:13,02160
Boiling Pt:59.4…61.5 °C
Melting Pt:–63 °C
Density:1.4832 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
ADR:6.1,III
REACH:01-2119486657-20
:GHS08
:GHS06
:GHS07
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826