Order Entry
Japan
ContactUsLinkComponent
Disodium hydrogen phosphate dodecahydrate 98.5-102.5% Ph. Eur.
Disodium hydrogen phosphate dodecahydrate 98.5-102.5% Ph. Eur.
Supplier:  VWR Chemicals
Certificates
undefined
Disodium hydrogen phosphate dodecahydrate 98.5-102.5% Ph. Eur.
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    98.5 - 102.5 %
  • Appearance:
    Conforms (see CoA/CoS)
  • Identification A:
    Passes test
  • Identification (PO4):
    Passes test
  • Identification (Na):
    Passes test
  • Appearance of solution:
    Passes test
  • Reducing substances:
    Passes test
  • Monosodium phosphate:
    ≤ 2.5 %
  • Cl (Chloride):
    ≤ 200 ppm
  • SO4 (Sulphate):
    ≤ 500 ppm
  • Fe (Iron):
    ≤ 20 ppm
  • Heavy metals (as Pb):
    ≤ 10 ppm
  • Loss on drying (130°C; 4 h):
    57.0 - 61.0 %
  • Residual solvents:
    Unlikely by manuf.process
  • As (Arsenic):
    ≤ 2 ppm
  • Cd (Cadmium):
    ≤ 1 ppm
  • Co (Cobalt):
    ≤ 1 ppm
  • Hg (Mercury):
    ≤ 2 ppm
  • Ni (Nickel):
    ≤ 1 ppm
  • Pb (Lead):
    ≤ 2 ppm
  • V (Vanadium):
    ≤ 4 ppm
  • Conforms to Ph.Eur.:
    Passes test

Specification Test Results

Frequently Bought Together