
Citric acid monohydrate ≥99%, powder, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:5949-29-1
:
Formula:C₆H₈O₇.H₂O
MDL Number:MFCD00149972
:210.14 g/mol
Merck Index:13,02350
Melting Pt:135…152 °C
Density:1.552 g/cm³ (20 °C)
Flash Pt:173.9 °C (closed cup)
EINECS:201-069-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 99 %
- Heavy metals (as Pb):Max. 20 ppm
- Ignition residue (SO4):Max. 0.05 %
- Cl (Chloride):Max. 20 ppm
- SO4 (Sulphate):Max. 100 ppm
- Fe (Iron):Max. 20 ppm
Specification Test Results
Frequently Bought Together