Order Entry
Japan
ContactUsLinkComponent
 
Copper ≥99.5%, turnings, AnalaR NORMAPUR® analytical reagent
 
undefined
Copper ≥99.5%, turnings, AnalaR NORMAPUR® analytical reagent
 

 

  • Assay:
    Min. 99.5 %
  • Insolubility in nitric acid:
    Max. 0.05 %
  • Ag (Silver):
    Max. 50 ppm
  • Fe + Al:
    Max. 0.025 %
  • Pb (Lead):
    Max. 0.05 %

 

Frequently Bought Together