VWR® Critical Print™ Cleanroom Pens and Markers
Catalog # 811-0125
Supplier: VWR Collection
Specifications
- Sterile:-
- Colour:Black
- Description:Cleanroom felt-tip marker
- Pk:12
Specifications
About this item
Manufactured with very low sodium to prevent sodium contamination.