VWR®, Measuring Cylinders, Tall Form, PP
Supplier: VWR Collection
Certificates
About this item
The 'nonwetting' polypropylene (PP) plastic surface of these single scale graduated cylinders eliminates formation of a concave meniscus and makes reading easier and more accurate. Chemically resistant, they are autoclavable at 121 °C (250 °F).
- Single metric scale has raised numerals for easy reading
- Flared top rims and large spouts facilitate pouring
- Octagonal-shaped bases prevent tipping and rolling
Frequently Bought Together