$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Japan
ContactUsLinkComponent
Destaining solution
Destaining solution Reag. Ph. Eur. 1012202
Destaining solution Reag. Ph. Eur. 1012202
Catalog # 87788.290
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Expand image
Destaining solution Reag. Ph. Eur. 1012202
Catalog # 87788.290
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Specifications
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
Specifications
About this item
Product Details
Documentation
:
Storage Temperature:
Ambient
UN:
2921
ADR:
8 (4.1),II
Product Details
Documentation