VWR®, Accessories for Refrigerated Circulators
Digital to analogue adapter, VWR Bath and refrigerated circulators, P and D models
Catalog # 461-0417
Supplier: VWR Collection
Jump to:
Product Details & Documents
For use with VWR® circulators with a programmable or digital controller when direct digital control is not required. Connects to the RS232 port and provides 10 mV/degree analogue signal for external monitoring or control.
Specifications
- Description:Digital to analogue adapter
- For:VWR Bath and refrigerated circulators, P and D models