Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, Sealing Mats for PCR Plates
VWR®, Sealing Mats for PCR Plates
Sealing mats, silicone, round wells
Catalog # 732-0589
   Low Manufacturing Impact|Sustainable Packaging
Supplier:   VWR Collection
VWR®, Sealing Mats for PCR Plates
Catalog # 732-0589
Supplier:  VWR Collection
   Low Manufacturing Impact|Sustainable Packaging
VWR®, Sealing Mats for PCR Plates
VWR®, Sealing Mats for PCR Plates
Sealing mats, silicone, round wells
Price
Inquire For Price
Log in for availability
1 * 5 items
Product Details & Documents
Transparent silicone mats fit most brands of 96-well PCR plates.
  • Sustainable design based on virgin resins
  • Can be cleaned and reused
  • Autoclavable at 121 °C
Designed to reduce evaporation when cycling 96-well plates.
Documents
Specifications
  • Description:
    Sealing mats, silicone, round wells
  • Seq:
    12
  • Cat. No.:
    732-0589
  • Environmentally Preferable:
    Low Manufacturing Impact|Sustainable Packaging