Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, Fire Blankets
VWR®, Fire Blankets
Supplier:   VWR Collection
VWR®, Fire Blankets
Supplier:  VWR Collection
Certificates
VWR®, Fire Blankets
VWR®, Fire Blankets
2 Options Available Below
Product Details & Documents
Made from a composite material including two plies of glass fibre matting and one layer of flame-retardant foil. For extinguishing and smothering relatively small fires. Environmentally friendly, these blankets contain no asbestos.
  • Heat resistant up to 550 °C
  • Safe, dust-free storage in a covered plastic box labelled in three languages (German, English, French)
  • Can be wall mounted
  • Unfolds quickly with two ripcords
BS EN 1869
Certifications: BS EN 1869
Documents
Product Family Options
Product Information
  • L×W
    Availability
    Price
  • 118-1105
    Fire blanket
    Fire blanket
    1200×1200 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 118-1106
    Fire blanket
    Fire blanket
    1200×1800 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items