VWR®, Blotting Pads, Grade 703
Ideal for use as wicks in protein and nucleic acid blotting. Provides a uniform flow of buffer through the gel to the transfer membrane in a blotting sandwich. Also suitable for use in removing gels from glass supports.
- Made of 100% cotton fibre
- Uniform and smooth
- Produced using ultra-pure water
- Contains no additives
- Catalog No:
- 732-0603
- 732-0605
- 732-0593
- 732-0596
- 732-0591
- Grade:
- 703
- 703
- 703
- 703
- 703
- Weight:
- 185 g/m²
- 185 g/m²
- 185 g/m²
- 185 g/m²
- 185 g/m²
- Thickness:
- 0,38 mm, 0,38 mm
- 0,38 mm, 0,38 mm
- 0,38 mm, 0,38 mm
- 0,38 mm, 0,38 mm
- 0,38 mm, 0,38 mm
- Herzberg flow rate:
- 250 s per 100 ml at 10 cm head of water
- 250 s per 100 ml at 10 cm head of water
- 250 s per 100 ml at 10 cm head of water
- 250 s per 100 ml at 10 cm head of water
- 250 s per 100 ml at 10 cm head of water
- Klemm wicking rate test:
- 626 s per 7,5 cm rise
- 626 s per 7,5 cm rise
- 626 s per 7,5 cm rise
- 626 s per 7,5 cm rise
- 626 s per 7,5 cm rise
Frequently Bought Together