Order Entry
Japan
ContactUsLinkComponent
VWR®, Beakers, Low Form
VWR®, Beakers, Low Form
Catalog # 213-1622
Supplier:  VWR Collection
Supplier #:
undefined
VWR®, Beakers, Low Form
Catalog # 213-1622
Supplier:  VWR Collection
Supplier #:
Log in for availability
Select an Option
47 mm
Select an Option
10 ml
Select an Option
50 ml
Select an Option
60 mm
Select an Option
02
Select an Option
12
About this item
PP, highly translucent.
  • Good chemical resistance
  • Resistant to heat up to +121 °C (for 20 minutes), prolonged use at +100 °C
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use. With spout and easy to read blue graduations.
ISO 7056
Certifications: ISO 7056
Frequently Bought Together