VWR®, Autoclavable Bags
Catalog # 129-0019
Supplier: VWR Collection
Specifications
- Capacity:45 L
- L×W:914×610 mm
- Material:PP
- Seq:09
- Thickness:37,5 µm
- Pk:500
Specifications
About this item
Sturdy, temperature resistant bags, ideal for disposing of hazardous waste that must first be autoclaved. Can also be used for autoclaving other products. Supplied complete with sealing wires. PP, 37,5 µm or 50 µm thick, transparent.
- Clear bag
- Autoclavable up to 135 °C
- Highly resistant to tears and impact