Order Entry
Japan
ContactUsLinkComponent
VWR®, Coulometry NewGen Reagent AG-FI for coulometric Karl Fischer titration
VWR®, Coulometry NewGen Reagent AG-FI for coulometric Karl Fischer titration
Catalog # 88425.260
Supplier:  VWR Chemicals
New Product
CAS Number:  
undefined
VWR®, Coulometry NewGen Reagent AG-FI for coulometric Karl Fischer titration
Catalog # 88425.260
Supplier:  VWR Chemicals
New Product
CAS Number:  
Specifications
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    NewGen Reagent AG-FI for coulometric Karl Fischer titration
  • Seq:
    12
Specifications
About this item
Anolyte for coulometric Karl Fischer titration, for cells with and without diaphragm, Methanol-based, free of Imidazole (free of CMR substances).
  • Free of imidazole
  • Free of CMR substances
Specification:
Suitability (for KF-Titration): complying
Water content: 5-50 ppm