Order Entry
Japan
ContactUsLinkComponent
VWR®, Coulometry NewGen Reagent A-FA for coulometric Karl Fischer titration
VWR®, Coulometry NewGen Reagent A-FA for coulometric Karl Fischer titration
Catalog # 88423.260
Supplier:  VWR Chemicals
New Product
CAS Number:  
undefined
VWR®, Coulometry NewGen Reagent A-FA for coulometric Karl Fischer titration
Catalog # 88423.260
Supplier:  VWR Chemicals
New Product
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    NewGen Reagent A-FA for coulometric Karl Fischer titration
  • Seq:
    10

Specifications

About this item

Anolyte for coulometric Karl Fischer titration of ketones and Li-ion battery electrolytes, preferred for cells with diaphragm, acetonitrile-based, free of alcohols (free of CMR substances)

  • free of alcohols
  • free of CMR substances
  • Recommended for Li-ion battery electrolytes

Specification:
Suitability (for KF-Titration): complying
Water content: max. 50 ppm