Order Entry
Japan
ContactUsLinkComponent
di-Sodium sulphide 0.5 mol/l in glycerine 85% 0.5 mol/l in glycerine 85% Reag. Ph. Eur. 1083901
Corrosive
Environment
di-Sodium sulphide 0.5 mol/l in glycerine 85% 0.5 mol/l in glycerine 85% Reag. Ph. Eur. 1083901
Catalog # 87940.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Environment
di-Sodium sulphide 0.5 mol/l in glycerine 85% 0.5 mol/l in glycerine 85% Reag. Ph. Eur. 1083901
Catalog # 87940.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :78.05 g/mol
Flash Pt:160 °C
Storage Temperature:Refrigerator
EINECS:215-211-5
UN:1760
ADR:8,III
 :GHS05
 :GHS09
CAS Number:1313-82-2
Formula:Na₂S
MDL Number:MFCD00003498