Order Entry
Japan
ContactUsLinkComponent
Potassium hydroxide 0.5 mol/l (0.5 N) in ethanol 10% Reag. Ph. Eur. 1070302
Potassium hydroxide 0.5 mol/l (0.5 N) in ethanol 10% Reag. Ph. Eur. 1070302
Catalog # 87905.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium hydroxide 0.5 mol/l (0.5 N) in ethanol 10% Reag. Ph. Eur. 1070302
Catalog # 87905.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :56.11 g/mol
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553