Order Entry
Japan
ContactUsLinkComponent
Perchloric acid 70% AR ACS, Analytical reagent (AR), Rankem®
Perchloric acid 70% AR ACS, Analytical reagent (AR), Rankem®
  RANKP0064
 :  Transene
 :  
Perchloric acid 70% AR ACS, Analytical reagent (AR), Rankem®
  RANKP0064
 :  Transene
 :  

 

  • Pack type:
    Amber coloured glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

 

 

  • Guarantee Analysis:
    .
  • Assay:
    70.00 - 72.00 %
  • Color:
    ≤ 10 APHA
  • Weight Per ml (at 20°C):
    1.67 - 1.69 g
  • Maximum Limits Of Impurities:
    Jun-03
  • Residue On Ignition:
    ≤ 0.003 %
  • Chlorate:
    <=0.0012%
  • Chloride (Cl):
    <= 0.001%
  • Free Chlorine:
    <=0.0005%
  • Nitrogen Compounds:
    <=0.001%
  • Phosphate And Silicate (Sio2):
    <=0.0005%
  • Sulphate (SO4):
    <=0.0005%
  • Heavy Metal (As Pb):
    <= 0.0001%
  • Copper(Cu):
    ≤ 0.10 ppm
  • Iron (Fe):
    ≤ 0.0001 %
  • Manganese (Mn):
    ≤ 0.02 ppm
  • Silver (Ag):
    ≤ 0.10 ppm
  • Cadmium (Cd):
    ≤ 0.05 ppm
  • Zinc (Zn):
    ≤ 0.50 ppm
  • Potassium (K):
    ≤ 0.002 %
  • Sodium (Na):
    ≤ 0.002 %

 

 

 :
 :100.46 g/mol
 :198.7 °C (1013 hPa)
 :–18 °C
 :1.67 g/cm³ (20 °C)
 :Ambient
 :231-512-4
 :1873
 :5.1,I
 :GHS03
 :GHS05
 :7601-90-3
 :HClO₄
 :MFCD00011325