Potassium hydrogen carbonate ≥99.5% (dry basis), powder USP, FCC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:298-14-6
:
Formula:KHCO₃
MDL Number:MFCD00011402
:100.12 g/mol
Merck Index:13,07691
Melting Pt:292 °C
Density:2,17 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-059-0
Specification Test Results
- Meets F.C.C. Requirements:99.5 - 101.5%
- Meets U.S.P Requirements:Passes Test
- GMP Manufactured Product:Passes Test
- Food GMP Manufactured Product:≤0.25%
- CAUTION: For Manufacturing, processing or repackaging:≤2.5%
- Bulk Pharmaceutical Chemical:≤10 ppm
- Bulk Food Chemical:99.5 - 101.5%
- USP - Assay (KHCO₃) (dried basis):99.5 - 101.5 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Loss on Drying:≤0.25 %
- USP - Normal Carbonate:≤2.5 %
- FCC - Assay (KHCO₃) (dried basis):99.5 - 101.5 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Carbonate:Passes Test
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Loss on Drying:≤0.25 %
- Solubility:Passes Test